siRNA Details
Gene Symbol: Myef2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013205.2 | NP_001013223.2 | 5, 6 | 490 | ||
XM_006234930.1 | XP_006234992.1 | 5, 6 | 616 | |||
XM_006234931.2 | XP_006234993.1 | 5, 6 | 616 | |||
XM_006234932.2 | XP_006234994.1 | 5, 6 | 616 | |||
XM_008762198.1 | XP_008760420.1 | 5, 6 | 616 |
