siRNA Details
Gene Symbol: Nop56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025732.1 | NP_001020903.1 | 11, 12 | 1481 | ||
GenBank mRNA | BC086568.1 | - | 8, 9 | 1159 | ||
BC099149.1 | - | 11, 12 | 1481 | 7366316 | ||
FQ232640.1 | - | 6, 7 | 727 |
