siRNA Details
Gene Symbol: Tbc1d20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004281.1 | NP_001004281.1 | 4 | 546 | ||
XM_008762348.1 | XP_008760570.1 | 4 | 695 | |||
GenBank mRNA | BC079383.1 | - | 4 | 546 | 7111239 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_024196, BC002196, AK006877, BC034504, AK076124, and others.
Gene Ontology Categories:
