siRNA Details
Gene Symbol: Dusp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108598.2 | NP_001102068.2 | 3, 4 | 338 | ||
NM_001244784.1 | NP_001231713.1 | 3, 4 | 338 | |||
GenBank mRNA | BC168211.1 | - | 3, 4 | 167 | 7135378 | |
CB576376.1 | - | 3, 4 | 215 | |||
CB585518.1 | - | 3, 4 | 338 | |||
CB736780.1 | - | 3, 4 | 417 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
