siRNA Details
Gene Symbol: Snta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100901.1 | NP_001094371.1 | 2 | 424 | ||
XM_006235326.2 | XP_006235388.1 | 2 | 463 | |||
GenBank mRNA | BC169101.1 | - | 2 | 424 | 5599966 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009228, U00677, BB625367, AK137379, AK037183, and others.
Gene Ontology Categories:
