siRNA Details
Gene Symbol: Map1lc3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199500.2 | NP_955794.1 | 4 | 461 | ||
GenBank mRNA | AY206668.1 | - | 4 | 409 | ||
BC086389.1 | - | 4 | 422 | 7302206 | ||
FM036735.1 | - | 3 | 246 | |||
FM046244.1 | - | 4 | 429 |

More Information
Other Species Targeted

Gene Ontology Categories:
