siRNA Details
Gene Symbol: Tp53inp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270947.1 | NP_001257876.1 | 2, 3 | 379 | ||
XM_003749598.3 | XP_003749646.1 | 2, 3 | 517 | |||
XM_006235327.2 | XP_006235389.1 | 2, 3 | 1091 | |||
XM_006235329.2 | XP_006235391.1 | 2, 3 | 1285 | |||
GenBank mRNA | AJ297794.1 | - | 2, 3 | 329 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
