siRNA Details
Gene Symbol: Procr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025733.2 | NP_001020904.1 | 3 | 642 | ||
GenBank mRNA | BC097380.1 | - | 3 | 613 | 7445753 | |
BC099115.1 | - | 3 | 642 | 7462042 | ||
FQ220150.1 | - | 3 | 681 | |||
FQ221148.1 | - | 3 | 649 | |||
FQ228484.1 | - | 3 | 647 | |||
FQ228710.1 | - | 3 | 649 | |||
FQ229445.1 | - | 3 | 679 | |||
FQ229483.1 | - | 3 | 649 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
