siRNA Details
Gene Symbol: Ptprt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108603.1 | NP_001102073.1 | 15 | 2331 | ||
XM_008762361.1 | XP_008760583.1 | 18 | 2753 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: AF162857.
Gene Ontology Categories:
