siRNA Details
Gene Symbol: Stx16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235693.2 | XP_006235755.1 | 5 | 1306 | ||
XM_006235694.2 | XP_006235756.1 | 4 | 1294 | |||
XM_006235695.2 | XP_006235757.1 | 5 | 1255 | |||
XM_006235696.2 | XP_006235758.1 | 4 | 1242 | |||
GenBank mRNA | BC168980.1 | - | 2 | 232 |

More Information
Other Species Targeted

Gene Ontology Categories:
