siRNA Details
Gene Symbol: Myt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108615.1 | NP_001102085.1 | 13 | 3188 | ||
XM_006235782.1 | XP_006235844.1 | 19 | 3005 | |||
XM_006235783.2 | XP_006235845.1 | 19 | 3239 | |||
XM_006235784.1 | XP_006235846.1 | 18 | 2936 | |||
XM_006235785.2 | XP_006235847.1 | 19 | 2956 | |||
XM_006235787.2 | XP_006235849.1 | 16 | 2826 |
