siRNA Details
Gene Symbol: Agap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235938.2 | XP_006236000.1 | 8, 9 | 1729 | ||
XM_006235939.2 | XP_006236001.1 | 8, 9 | 1728 | |||
XM_006235940.2 | XP_006236002.1 | 8, 9 | 1727 | |||
XM_006235941.2 | XP_006236003.1 | 8, 9 | 1418 | |||
XM_006235942.2 | XP_006236004.1 | 8, 9 | 1417 | |||
XM_008762650.1 | XP_008760872.1 | 8, 9 | 1033 | |||
GenBank mRNA | KF447874.1 | - | 8, 9 | 1123 |

More Information
Other Species Targeted

Gene Ontology Categories:
