siRNA Details
Gene Symbol: Abcb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007796.1 | NP_001007797.1 | 14 | 1642 | ||
XM_006235946.2 | XP_006236008.1 | 13 | 1438 | |||
GenBank mRNA | BC085781.1 | - | 14 | 1685 | 7107433 | |
DQ233644.1 | - | 14 | 1702 | |||
FQ216748.1 | - | 1 | 266 |

More Information
Other Species Targeted

Gene Ontology Categories:
