siRNA Details
Gene Symbol: Klhl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012187.1 | NP_001012187.1 | 2, 3 | 257 | ||
GenBank mRNA | BC083903.1 | - | 2, 3 | 257 | 7133606 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Klhl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012187.1 | NP_001012187.1 | 2, 3 | 257 | ||
GenBank mRNA | BC083903.1 | - | 2, 3 | 257 | 7133606 |