siRNA Details
Gene Symbol: Krit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108618.1 | NP_001102088.1 | 6 | 977 | ||
GenBank mRNA | BC168923.1 | - | 6 | 984 | 7105532 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_030675, NM_001170552, AK039264, AF306509, BC085252, and others.
Gene Ontology Categories:
