siRNA Details
Gene Symbol: Tspan12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015026.1 | NP_001015026.1 | 5 | 694 | ||
XM_008762784.1 | XP_008761006.1 | 6 | 616 | |||
XM_008762785.1 | XP_008761007.1 | 6 | 545 | |||
XM_008762786.1 | XP_008761008.1 | 6 | 544 | |||
XM_008762787.1 | XP_008761009.1 | 6 | 545 | |||
XM_008762788.1 | XP_008761010.1 | 5 | 564 | |||
XM_008762789.1 | XP_008761011.1 | 5 | 827 | |||
XM_008762790.1 | XP_008761012.1 | 5 | 827 | |||
GenBank mRNA | BC092623.1 | - | 5 | 694 | 7313631 |
