siRNA Details
Gene Symbol: Impdh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108619.1 | NP_001102089.1 | 14 | 1602 | ||
XM_006236208.1 | XP_006236270.1 | 14 | 1824 | |||
XM_006236210.1 | XP_006236272.1 | 14 | 1824 | |||
XM_008762791.1 | XP_008761013.1 | 14 | 1827 | |||
XM_008762792.1 | XP_008761014.1 | 13 | 1752 | |||
XM_008762793.1 | XP_008761015.1 | 14 | 1827 | |||
XM_008762794.1 | XP_008761016.1 | 13 | 1752 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011829, AK047537, U00978, AK089383, AK171139, and others.
Gene Ontology Categories:
