siRNA Details
Gene Symbol: Cul1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108627.2 | NP_001102097.1 | 9 | 1228 | ||
XM_006236409.1 | XP_006236471.1 | 8 | 1136 | |||
XM_006236410.2 | XP_006236472.1 | 8 | 1138 | |||
XM_006236411.1 | XP_006236473.1 | 8 | 1136 | |||
GenBank mRNA | BC161932.1 | - | 9 | 1228 | 9021933 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_012042, AK147004, AF136441, AF083216, AK209075, and others.
Gene Ontology Categories:
