siRNA Details
Gene Symbol: Mpp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134982.1 | NP_001128454.1 | 10 | 1432 | ||
XM_006236496.2 | XP_006236558.1 | 11 | 1493 | |||
XM_006236497.2 | XP_006236559.1 | 11 | 1559 | |||
XM_008762895.1 | XP_008761117.1 | 11 | 1974 | |||
XM_008762897.1 | XP_008761119.1 | 11 | 1513 | |||
XM_008762898.1 | XP_008761120.1 | 10 | 1931 | |||
GenBank mRNA | BC169025.1 | - | 10 | 1432 | 9027949 |
