siRNA Details
Gene Symbol: Mpp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134982.1 | NP_001128454.1 | 10, 11 | 1499 | ||
XM_006236496.2 | XP_006236558.1 | 11, 12 | 1560 | |||
XM_006236497.2 | XP_006236559.1 | 11, 12 | 1626 | |||
XM_008762895.1 | XP_008761117.1 | 11, 12 | 2041 | |||
XM_008762897.1 | XP_008761119.1 | 11, 12 | 1580 | |||
XM_008762898.1 | XP_008761120.1 | 10, 11 | 1998 | |||
GenBank mRNA | BC169025.1 | - | 10, 11 | 1499 | 9027949 |
