siRNA Details
Gene Symbol: Znrf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108628.1 | NP_001102098.1 | 3, 4 | 696 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_199143, BC104316, BC104317, BC125242, AF513708.
Gene Ontology Categories:
