siRNA Details
Gene Symbol: Vopp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108630.1 | NP_001102100.1 | 2, 3 | 271 | ||
XM_006236578.2 | XP_006236640.1 | 2, 3 | 420 | |||
GenBank mRNA | BC168709.1 | - | 2, 3 | 264 | 8363599 |

More Information
Other Species Targeted

Gene Ontology Categories:
