siRNA Details
Gene Symbol: Mrpl53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108635.2 | NP_001102105.1 | 1 | 53 | ||
GenBank mRNA | BC166928.1 | - | 1 | 53 | 6920305 |
Gene Symbol: LOC103692169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763084.1 | XP_008761306.1 | 1 | 323 |

More Information
Other Species Targeted

