siRNA Details
Gene Symbol: Isy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014188.1 | NP_001014210.1 | 8 | 550 | ||
GenBank mRNA | BC079119.1 | - | 8 | 550 | 7126525 | |
FQ218201.1 | - | 8 | 601 |

More Information
Other Species Targeted

Gene Ontology Categories:
