siRNA Details
Gene Symbol: Isy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014188.1 | NP_001014210.1 | 10 | 797 | ||
GenBank mRNA | BC079119.1 | - | 10 | 797 | 7126525 | |
FQ218201.1 | - | 9 | 1584 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Isy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014188.1 | NP_001014210.1 | 10 | 797 | ||
GenBank mRNA | BC079119.1 | - | 10 | 797 | 7126525 | |
FQ218201.1 | - | 9 | 1584 |