siRNA Details
Gene Symbol: Rad18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077673.1 | NP_001071141.1 | 7 | 848 | ||
XM_006237036.1 | XP_006237098.1 | 7 | 877 | |||
XM_006237037.1 | XP_006237099.1 | 7 | 877 | |||
GenBank mRNA | BC127510.1 | - | 7 | 848 | 8366167 | |
EF395817.1 | - | 818 |
