siRNA Details
Gene Symbol: Tada3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025734.1 | NP_001020905.1 | 9 | 1515 | ||
XM_006237038.1 | XP_006237100.1 | 10 | 1982 | |||
XM_006237040.1 | XP_006237102.1 | 10 | 1808 | |||
XM_006237042.1 | XP_006237104.1 | 9 | 1754 | |||
XM_006237043.1 | XP_006237105.1 | 9 | 1796 | |||
GenBank mRNA | BC097414.1 | - | 9 | 1515 | 7454313 |
