siRNA Details
Gene Symbol: Il17re | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004091.1 | NP_001004091.1 | 8, 9 | 929 | ||
GenBank mRNA | BC078742.1 | - | 8, 9 | 929 | 7123764 |

More Information
Other Species Targeted

Gene Ontology Categories:
