siRNA Details
Gene Symbol: Rassf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024275.1 | NP_001019446.1 | 2 | 123 | ||
GenBank mRNA | BC093620.1 | - | 2 | 123 | 7315142 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
