siRNA Details
Gene Symbol: Mical3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191085.1 | NP_001178014.1 | 4 | 717 | ||
GenBank mRNA | BC083659.1 | - | 4 | 718 | ||
BC160830.1 | - | 4 | 920 | 7453653 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mical3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191085.1 | NP_001178014.1 | 4 | 717 | ||
GenBank mRNA | BC083659.1 | - | 4 | 718 | ||
BC160830.1 | - | 4 | 920 | 7453653 |