siRNA Details
Gene Symbol: Mical3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191085.1 | NP_001178014.1 | 6, 7 | 1058 | ||
GenBank mRNA | BC083659.1 | - | 6, 7 | 1059 | ||
BC160830.1 | - | 6, 7 | 1261 | 7453653 |
