siRNA Details
Gene Symbol: Clec4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005891.1 | NP_001005891.1 | 2 | 184 | ||
XM_006237321.2 | XP_006237383.1 | 2 | 393 | |||
GenBank mRNA | AY397671.1 | - | 2 | 184 | ||
AY494062.1 | - | 2 | 185 | |||
FQ230051.1 | - | 2 | 279 | |||
FQ234061.1 | - | 2 | 309 |

More Information
Other Species Targeted

