siRNA Details
Gene Symbol: Clec4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003707.1 | NP_001003707.1 | 2, 3 | 163 | ||
XM_006237322.2 | XP_006237384.1 | 2, 3 | 348 | |||
GenBank mRNA | AY339882.1 | - | 2, 3 | 163 | ||
AY494065.1 | - | 2, 3 | 163 |
