siRNA Details
Gene Symbol: Ndufa9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100752.1 | NP_001094222.1 | 6 | 628 | ||
XM_006237500.1 | XP_006237562.1 | 6 | 711 | |||
GenBank mRNA | BC091192.1 | - | 6 | 594 | ||
BC168777.1 | - | 6 | 628 | 6922152 | ||
FQ216233.1 | - | 6 | 628 | |||
FQ216540.1 | - | 6 | 628 | |||
FQ216671.1 | - | 6 | 618 | |||
FQ216764.1 | - | 6 | 618 | |||
FQ232722.1 | - | 6 | 626 | |||
FQ233877.1 | - | 6 | 696 |
