siRNA Details
Gene Symbol: Loh12cr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108650.2 | NP_001102120.1 | 2 | 155 | ||
XM_006237528.2 | XP_006237590.1 | 2 | 888 | |||
XM_008763394.1 | XP_008761616.1 | 2 | 886 | |||
XM_008763395.1 | XP_008761617.1 | 2 | 886 | |||
XM_008763396.1 | XP_008761618.1 | 2 | 888 | |||
GenBank mRNA | BC160872.1 | - | 2 | 155 | 7365768 | |
FQ211769.1 | - | 2 | 250 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
