siRNA Details
Gene Symbol: Hebp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237529.1 | XP_006237591.1 | 3, 4 | 508 | ||
GenBank mRNA | BC168221.1 | - | 3, 4 | 457 | 7323058 | |
FQ220508.1 | - | 3, 4 | 499 | |||
FQ229464.1 | - | 3, 4 | 499 | |||
FQ235087.1 | - | 3, 4 | 468 |
