siRNA Details
Gene Symbol: RGD1306151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108652.1 | NP_001102122.1 | 9, 10 | 1399 | ||
XM_006237532.1 | XP_006237594.1 | 9, 10 | 1491 | |||
XM_008763398.1 | XP_008761620.1 | 9, 10 | 2307 | |||
XM_008763399.1 | XP_008761621.1 | 9, 10 | 2276 |
