siRNA Details
Gene Symbol: Arhgdib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009600.1 | NP_001009600.1 | 4 | 365 | ||
XM_006237561.2 | XP_006237623.1 | 4 | 389 | |||
XM_006237562.2 | XP_006237624.1 | 4 | 520 | |||
XM_006237563.2 | XP_006237625.1 | 5 | 451 | |||
XM_006237564.2 | XP_006237626.1 | 4 | 480 | |||
XM_006237565.2 | XP_006237627.1 | 4 | 665 | |||
GenBank mRNA | BC088209.1 | - | 4 | 365 | 7382227 | |
FQ225418.1 | - | 4 | 339 | |||
FQ233135.1 | - | 4 | 346 | |||
FQ234136.1 | - | 4 | 360 |

More Information
Other Species Targeted

Gene Ontology Categories:
