siRNA Details
Gene Symbol: Adhfe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025423.1 | NP_001020594.1 | 6 | 471 | ||
XM_008763507.1 | XP_008761729.1 | 6 | 852 | |||
XM_008763508.1 | XP_008761730.1 | 7 | 519 | |||
XM_008763509.1 | XP_008761731.1 | 4 | 273 | |||
XM_008763510.1 | XP_008761732.1 | 4 | 650 | |||
GenBank mRNA | BC085821.1 | - | 6 | 474 | ||
BC097945.1 | - | 6 | 471 | 7323001 |
