siRNA Details
Gene Symbol: Tcea1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025735.1 | NP_001020906.1 | 2 | 175 | ||
XM_006237816.2 | XP_006237878.1 | 2 | 236 | |||
XM_008763546.1 | XP_008761768.1 | 2 | 63 | |||
GenBank mRNA | BC099141.1 | - | 2 | 175 | 7460791 | |
FQ227501.1 | - | 2 | 147 |
Gene Symbol: LOC498453 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251720.2 | XP_006251782.1 | 1 | 229 |

More Information
Other Species Targeted

Gene Ontology Categories:
