siRNA Details
Gene Symbol: Tox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108654.1 | NP_001102124.1 | 2, 3 | 313 | ||
XM_006237844.2 | XP_006237906.1 | 2, 3 | 672 | |||
XM_008763547.1 | XP_008761769.1 | 1, 2 | 418 |

More Information
Other Species Targeted

Gene Ontology Categories:
