siRNA Details
Gene Symbol: Runx1t1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237901.2 | XP_006237963.1 | 2, 3 | 489 | ||
XM_008763549.1 | XP_008761771.1 | 1, 2 | 5652 | |||
XM_008763550.1 | XP_008761772.1 | 2, 3 | 185 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001111027, BY140482, X79989.
Gene Ontology Categories:
