siRNA Details
Gene Symbol: Runx1t1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237901.2 | XP_006237963.1 | 2 | 479 | ||
XM_008763549.1 | XP_008761771.1 | 1 | 5642 | |||
XM_008763550.1 | XP_008761772.1 | 2 | 175 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
