siRNA Details
Gene Symbol: Ripk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191865.1 | NP_001178794.1 | 4 | 671 | ||
GenBank mRNA | BC107468.1 | - | 2 | 102 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ripk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191865.1 | NP_001178794.1 | 4 | 671 | ||
GenBank mRNA | BC107468.1 | - | 2 | 102 |