siRNA Details
Gene Symbol: Ndufaf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198783.1 | NP_942078.1 | 3 | 397 | ||
GenBank mRNA | AF283900.1 | - | 3 | 397 | ||
AF283901.1 | - | 2 | 660 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
