siRNA Details
Gene Symbol: Anks6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015028.2 | NP_001015028.2 | 12, 13 | 2488 | ||
XM_008763699.1 | XP_008761921.1 | 12, 13 | 2359 | |||
XM_008763700.1 | XP_008761922.1 | 13 | 3041 | |||
GenBank mRNA | AY661303.1 | - | 12, 13 | 2488 |
