siRNA Details
Gene Symbol: Msantd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108664.1 | NP_001102134.1 | 2 | 363 | ||
XM_008763701.1 | XP_008761923.1 | 1 | 604 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_028137, NM_001145925, NM_001145924, AK017795, AK013199, and others.
Gene Ontology Categories:
