siRNA Details
Gene Symbol: Fktn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108667.1 | NP_001102137.1 | 5 | 950 | ||
XM_006238166.2 | XP_006238228.1 | 7 | 1057 | |||
XM_006238168.2 | XP_006238230.1 | 6 | 984 | |||
XM_006238169.1 | XP_006238231.1 | 5 | 867 | |||
XM_008763712.1 | XP_008761934.1 | 6 | 1077 | |||
XM_008763713.1 | XP_008761935.1 | 7 | 1051 | |||
XM_008763714.1 | XP_008761936.1 | 6 | 985 | |||
XM_008763715.1 | XP_008761937.1 | 6 | 984 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
