siRNA Details
Gene Symbol: Fktn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108667.1 | NP_001102137.1 | 8 | 1267 | ||
XM_006238166.2 | XP_006238228.1 | 10 | 1374 | |||
XM_006238168.2 | XP_006238230.1 | 9 | 1301 | |||
XM_006238169.1 | XP_006238231.1 | 8 | 1184 | |||
XM_008763712.1 | XP_008761934.1 | 9 | 1394 | |||
XM_008763713.1 | XP_008761935.1 | 10 | 1368 | |||
XM_008763715.1 | XP_008761937.1 | 9 | 1301 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
