siRNA Details
Gene Symbol: Fktn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108667.1 | NP_001102137.1 | 7, 8 | 1248 | ||
XM_006238166.2 | XP_006238228.1 | 9, 10 | 1355 | |||
XM_006238168.2 | XP_006238230.1 | 8, 9 | 1282 | |||
XM_006238169.1 | XP_006238231.1 | 7, 8 | 1165 | |||
XM_008763712.1 | XP_008761934.1 | 8, 9 | 1375 | |||
XM_008763713.1 | XP_008761935.1 | 9, 10 | 1349 | |||
XM_008763715.1 | XP_008761937.1 | 8, 9 | 1282 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
